siRNA Details
| Gene Symbol: Tbx5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011537.3 | NP_035667.1 | 4 | 991 | ||
| GenBank mRNA | AF140427.1 | - | 4 | 756 | ||
| BC090639.1 | - | 4 | 995 | 30548234 | ||
| U57330.1 | - | 2 | 152 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tbx5| Gene Symbol: Tbx5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011537.3 | NP_035667.1 | 4 | 991 | ||
| GenBank mRNA | AF140427.1 | - | 4 | 756 | ||
| BC090639.1 | - | 4 | 995 | 30548234 | ||
| U57330.1 | - | 2 | 152 | |||