siRNA Details
| Gene Symbol: Tcf12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001253862.1 | NP_001240791.1 | 15 | 1581 | ||
| NM_001253863.1 | NP_001240792.1 | 9 | 1032 | |||
| NM_001253864.1 | NP_001240793.1 | 8 | 960 | |||
| NM_001253865.1 | NP_001240794.1 | 10 | 1207 | |||
| NM_011544.3 | NP_035674.2 | 16 | 1710 | |||
| GenBank mRNA | AK132002.1 | - | 9 | 1020 | ||
| AK133958.1 | - | 9 | 1020 | |||
| AK134037.1 | - | 15 | 1582 | |||
| AK135376.1 | - | 8 | 928 | |||
| BC037097.1 | - | 10 | 1195 | 5345693 | ||
| M97635.1 | - | 16 | 1681 | |||
| M97636.1 | - | 1 | 1685 | |||
| X64840.1 | - | 16 | 1510 | |||

Tcf12
Genomic Map