siRNA Details
| Gene Symbol: RAB40B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006822.2 | NP_006813.1 | 6 | 1190 | ||
| XM_005256334.4 | XP_005256391.1 | 6 | 1393 | |||
| XM_006722271.3 | XP_006722334.1 | 6 | 959 | |||
| XM_011523528.2 | XP_011521830.1 | 6 | 1312 | |||
| XM_017024042.1 | XP_016879531.1 | 7 | 1825 | |||
| XM_017024043.1 | XP_016879532.1 | 6 | 1302 | |||
| GenBank mRNA | AK095782.1 | - | 1 | 1954 | ||
| AK124376.1 | - | 2 | 1806 | |||
| BC018039.1 | - | 6 | 1190 | 4801271 | ||
| U05227.1 | - | 6 | 1109 | |||

RAB40B
Genomic Map