siRNA Details
| Gene Symbol: Tcf12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001253862.1 | NP_001240791.1 | 9 | 823 | ||
| NM_001253863.1 | NP_001240792.1 | 2 | 202 | |||
| NM_001253864.1 | NP_001240793.1 | 2 | 202 | |||
| NM_001253865.1 | NP_001240794.1 | 4 | 449 | |||
| NM_011544.3 | NP_035674.2 | 9 | 880 | |||
| GenBank mRNA | AK078415.1 | - | 2 | 195 | ||
| AK132002.1 | - | 2 | 187 | |||
| AK133958.1 | - | 2 | 190 | |||
| AK134037.1 | - | 9 | 824 | |||
| AK135376.1 | - | 2 | 170 | |||
| BC037097.1 | - | 4 | 437 | 5345693 | ||
| CJ164494.1 | - | 2 | 204 | |||
| M97635.1 | - | 9 | 851 | |||
| X64840.1 | - | 9 | 680 | |||

Tcf12
Genomic Map