siRNA Details
| Gene Symbol: Tdgf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011562.2 | NP_035692.2 | 3 | 434 | ||
| GenBank mRNA | AK133730.1 | - | 3 | 372 | ||
| AK163969.1 | - | 3 | 371 | |||
| BC052646.1 | - | 3 | 376 | 30029798 | ||
| M87321.1 | - | 3 | 343 | |||

Tdgf1
Genomic Map