siRNA Details
| Gene Symbol: Tex264 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081654.2 | NP_001075123.1 | 6 | 1155 | ||
| NM_001286498.1 | NP_001273427.1 | 6 | 1355 | |||
| NM_011573.3 | NP_035703.2 | 6 | 1610 | |||
| GenBank mRNA | AK003113.1 | - | 7 | 1116 | ||
| AK009326.1 | - | 6 | 1461 | |||
| AK159046.1 | - | 6 | 1861 | |||
| BC002248.1 | - | 6 | 1119 | 3493141 | ||
| BC006608.1 | - | 6 | 1093 | 3157453 | ||

Tex264
Genomic Map