siRNA Details
| Gene Symbol: Cirh1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011574.2 | NP_035704.2 | 7 | 854 | ||
| GenBank mRNA | AK144455.1 | - | 7 | 839 | ||
| AK146263.1 | - | 7 | 830 | |||
| AK150862.1 | - | 7 | 829 | |||
| AK168470.1 | - | 7 | 804 | |||
| AK173318.1 | - | 6 | 795 | |||
| AK179441.1 | - | 2 | 86 | |||
| AK187526.1 | - | 3 | 303 | |||
| AK195627.1 | - | 3 | 308 | |||
| BC027399.1 | - | 3 | 353 | 4948207 | ||
| BC068113.1 | - | 7 | 843 | 5707865 | ||

Cirh1a
Genomic Map