siRNA Details
| Gene Symbol: Tfpi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177319.1 | NP_001170790.1 | 4 | 444 | ||
| NM_001177320.1 | NP_001170791.1 | 4 | 410 | |||
| NM_011576.1 | NP_035706.1 | 4 | 607 | |||
| GenBank mRNA | AF004833.1 | - | 3 | 376 | ||
| AF016313.1 | - | 2 | 251 | |||
| AK028356.1 | - | 4 | 607 | |||
| AK034752.1 | - | 4 | 411 | |||
| AK167522.1 | - | 4 | 469 | |||
| BC036146.1 | - | 4 | 444 | 4975683 | ||

Tfpi
Genomic Map