siRNA Details
Gene Symbol: Tgtp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011579.3 | NP_035709.3 | 2 | 818 | ||
GenBank mRNA | AK089836.1 | - | 2 | 820 | ||
AK163978.1 | - | 2 | 819 | |||
AK172473.1 | - | 2 | 818 | |||
BC034256.1 | - | 2 | 783 | 3662733 | ||
BC085259.1 | - | 789 | 5389279 | |||
BC093522.1 | - | 2 | 800 | 3988138 | ||
CT010183.1 | - | 1 | 557 | |||
U15636.1 | - | 2 | 662 |
Gene Symbol: Tgtp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145164.1 | NP_001138636.1 | 2 | 825 | ||
GenBank mRNA | AK088858.1 | - | 2 | 819 | ||
AK172580.1 | - | 2 | 818 | |||
L38444.1 | - | 2 | 656 |
