siRNA Details
| Gene Symbol: Thbs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011581.3 | NP_035711.2 | 3 | 792 | ||
| GenBank mRNA | AK132508.1 | - | 3 | 623 | ||
| AK142552.1 | - | 3 | 766 | |||
| BC053702.1 | - | 3 | 594 | 30062369 | ||
| L07803.1 | - | 3 | 620 | |||
| M64866.1 | - | 3 | 620 | |||

Thbs2
Genomic Map