siRNA Details
Gene Symbol: Tia1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011585.4 | NP_035715.1 | 4, 5 | 553 | ||
GenBank mRNA | AK088486.1 | - | 4, 5 | 489 | ||
AK144222.1 | - | 4, 5 | 506 | |||
AK185472.1 | - | 2, 3 | 71 | |||
U00689.1 | - | 4, 5 | 494 | |||
U55862.1 | - | 4, 5 | 361 |
