siRNA Details
| Gene Symbol: Tia1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011585.4 | NP_035715.1 | 4, 5 | 561 | ||
| GenBank mRNA | AK088486.1 | - | 4, 5 | 497 | ||
| AK144222.1 | - | 4, 5 | 514 | |||
| AK185472.1 | - | 2, 3 | 79 | |||
| U00689.1 | - | 4, 5 | 502 | |||
| U55862.1 | - | 4, 5 | 369 | |||

Tia1
Genomic Map