siRNA Details
| Gene Symbol: Timm17a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011590.2 | NP_035720.1 | 4, 5 | 339 | ||
| GenBank mRNA | AF106620.1 | - | 4, 5 | 321 | ||
| AK003114.1 | - | 4, 5 | 346 | |||
| AK003129.1 | - | 4, 5 | 340 | |||
| BC010830.1 | - | 4, 5 | 327 | 4220707 | ||
| BC098216.1 | - | 4, 5 | 336 | 1527710 | ||

Timm17a
Genomic Map