siRNA Details
| Gene Symbol: Timm17a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011590.2 | NP_035720.1 | 6 | 810 | ||
| GenBank mRNA | AK003129.1 | - | 6 | 811 | ||
| BC010830.1 | - | 6 | 798 | 4220707 | ||
| BC098216.1 | - | 6 | 807 | 1527710 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Timm17a| Gene Symbol: Timm17a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011590.2 | NP_035720.1 | 6 | 810 | ||
| GenBank mRNA | AK003129.1 | - | 6 | 811 | ||
| BC010830.1 | - | 6 | 798 | 4220707 | ||
| BC098216.1 | - | 6 | 807 | 1527710 | ||