siRNA Details
| Gene Symbol: Tmpo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080129.2 | NP_001073598.1 | 3 | 764 | ||
| NM_001080130.2 | NP_001073599.1 | 3 | 764 | |||
| NM_001080131.2 | NP_001073600.1 | 3 | 764 | |||
| NM_001080132.2 | NP_001073601.1 | 3 | 764 | |||
| NM_001283048.1 | NP_001269977.1 | 3 | 447 | |||
| NM_011605.3 | NP_035735.2 | 3 | 764 | |||
| GenBank mRNA | AK011988.1 | - | 3 | 257 | ||
| AK011992.1 | - | 3 | 257 | |||
| AK141845.1 | - | 3 | 714 | |||
| AK144642.1 | - | 3 | 724 | |||
| AK146903.1 | - | 3 | 731 | |||
| AK150305.1 | - | 3 | 712 | |||
| AK150515.1 | - | 3 | 699 | |||
| AK151936.1 | - | 3 | 716 | |||
| AK152737.1 | - | 3 | 716 | |||
| AK153281.1 | - | 3 | 699 | |||
| AK156206.1 | - | 3 | 714 | |||
| AK159161.1 | - | 3 | 360 | |||
| AK165282.1 | - | 3 | 714 | |||
| AK165851.1 | - | 3 | 721 | |||
| AK192607.1 | - | 3 | 203 | |||
| BC141062.1 | - | 3 | 482 | 9055674 | ||
| BE852115.1 | - | 3 | 205 | |||
| U39073.1 | - | 3 | 667 | |||
| U39074.1 | - | 3 | 695 | |||
| U39075.1 | - | 3 | 695 | |||
| U39076.1 | - | 3 | 695 | |||
| U39077.1 | - | 3 | 695 | |||
| U39078.1 | - | 3 | 678 | |||

Tmpo
Genomic Map