siRNA Details
| Gene Symbol: Tnc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011607.3 | NP_035737.2 | 7 | 2820 | ||
| GenBank mRNA | BC117979.1 | - | 6 | 2544 | 40099589 | |
| BC117980.1 | - | 6 | 2544 | 40099596 | ||
| CB521403.1 | - | 2 | 79 | |||
| CN692948.1 | - | 4 | 375 | |||
| D90343.1 | - | 7 | 2621 | |||
| X56304.1 | - | 6 | 2554 | |||

Tnc
Genomic Map