siRNA Details
| Gene Symbol: Tnfsf12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011614.3 | NP_035744.1 | 7 | 1074 | ||
| GenBank mRNA | AF030100.2 | - | 7 | 730 | ||
| AK020909.1 | - | 4 | 413 | |||
| AK170297.1 | - | 7 | 974 | |||
| AK171014.1 | - | 7 | 974 | |||
| CB204861.1 | - | 2 | 352 | |||

Tnfsf12
Genomic Map