siRNA Details
| Gene Symbol: Tnnt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163664.1 | NP_001157136.1 | 13 | 447 | ||
| NM_001163665.1 | NP_001157137.1 | 13 | 447 | |||
| NM_001163666.1 | NP_001157138.1 | 11 | 420 | |||
| NM_001163667.1 | NP_001157139.1 | 12 | 435 | |||
| NM_001163668.1 | NP_001157140.1 | 11 | 396 | |||
| NM_001163669.1 | NP_001157141.1 | 12 | 408 | |||
| NM_001163670.1 | NP_001157142.1 | 9 | 363 | |||
| NM_011620.3 | NP_035750.2 | 10 | 375 | |||
| GenBank mRNA | AA028290.1 | - | 12 | 388 | ||
| AA754928.1 | - | 10 | 351 | |||
| AA771596.1 | - | 11 | 373 | |||
| AI173874.1 | - | 11 | 412 | |||
| AI574395.1 | - | 13 | 441 | |||
| AI746973.1 | - | 11 | 415 | |||
| AK003147.1 | - | 9 | 330 | |||
| BC003747.1 | - | 13 | 443 | 3594256 | ||
| CB196352.1 | - | 3 | 199 | |||
| CK791812.1 | - | 2 | 85 | |||
| DV049834.1 | - | 10 | 422 | |||
| L48988.1 | - | 7 | 266 | |||
| L48989.1 | - | 7 | 266 | |||
| L48990.1 | - | 10 | 311 | |||
| L48991.1 | - | 7 | 293 | |||
| L48992.1 | - | 9 | 299 | |||
| L48993.1 | - | 9 | 299 | |||
| L49018.1 | - | 7 | 293 | |||
| L49466.1 | - | 12 | 435 | |||
| L49467.1 | - | 13 | 447 | |||
| L49468.1 | - | 11 | 420 | |||
| L49469.1 | - | 11 | 420 | |||
| L49470.1 | - | 12 | 408 | |||
| L49471.1 | - | 11 | 417 | |||
| L49472.1 | - | 11 | 396 | |||
| U77779.1 | - | 10 | 368 | |||

Tnnt3
Genomic Map