siRNA Details
| Gene Symbol: Tom1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136259.1 | NP_001129731.1 | 8 | 962 | ||
| NM_011622.3 | NP_035752.1 | 8 | 962 | |||
| GenBank mRNA | AJ006972.1 | - | 8 | 939 | ||
| AK028398.1 | - | 8 | 903 | |||
| AK051245.1 | - | 8 | 944 | |||
| AK150143.1 | - | 8 | 912 | |||
| AK160403.1 | - | 8 | 908 | |||
| BC021633.1 | - | 8 | 925 | 4208226 | ||
| BC093520.1 | - | 8 | 890 | 2866013 | ||
| BC157905.1 | - | 8 | 923 | 9088106 | ||
| BC158042.1 | - | 8 | 923 | 9007343 | ||

Tom1
Genomic Map