siRNA Details
| Gene Symbol: Tom1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136259.1 | NP_001129731.1 | 10 | 1049 | ||
| NM_011622.3 | NP_035752.1 | 10 | 1049 | |||
| GenBank mRNA | AJ006972.1 | - | 10 | 1026 | ||
| AK028398.1 | - | 10 | 990 | |||
| AK051245.1 | - | 10 | 1031 | |||
| AK150143.1 | - | 10 | 999 | |||
| AK160403.1 | - | 10 | 995 | |||
| AK189274.1 | - | 3 | 81 | |||
| BC021633.1 | - | 10 | 1012 | 4208226 | ||
| BC093520.1 | - | 10 | 977 | 2866013 | ||
| BC157905.1 | - | 10 | 1010 | 9088106 | ||
| BC158042.1 | - | 10 | 1010 | 9007343 | ||

Tom1
Genomic Map