siRNA Details
| Gene Symbol: GLIPR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006851.2 | NP_006842.2 | 1 | 297 | ||
| XM_011537775.2 | XP_011536077.1 | 1 | 276 | |||
| GenBank mRNA | AF400440.1 | - | 1 | 177 | ||
| AK304847.1 | - | 1 | 277 | |||
| AK313449.1 | - | 1 | 163 | |||
| BC012510.1 | - | 1 | 213 | 4453841 | ||
| CB123261.1 | - | 1 | 350 | |||
| EF667354.1 | - | 1 | 149 | |||
| U16307.3 | - | 1 | 233 | |||
| X91911.1 | - | 1 | 277 | |||

GLIPR1
Genomic Map