siRNA Details
| Gene Symbol: Tsn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011650.3 | NP_035780.1 | 5 | 453 | ||
| GenBank mRNA | AF234179.1 | - | 5 | 453 | ||
| AK010737.1 | - | 5 | 451 | |||
| AK150529.1 | - | 5 | 502 | |||
| AK165598.1 | - | 5 | 493 | |||
| AK219078.1 | - | 2 | 114 | |||
| BC004615.1 | - | 5 | 499 | 3585366 | ||
| X81464.1 | - | 5 | 453 | |||

Tsn
Genomic Map