siRNA Details
| Gene Symbol: Tuft1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001293728.1 | NP_001280657.1 | 11 | 1047 | ||
| NM_011656.3 | NP_035786.1 | 12 | 1122 | |||
| GenBank mRNA | AF047704.1 | - | 12 | 1056 | ||
| AK132488.1 | - | 12 | 1054 | |||
| AK171705.1 | - | 11 | 1012 | |||
| BC019213.1 | - | 12 | 1053 | 5064658 | ||

Tuft1
Genomic Map