siRNA Details
| Gene Symbol: Tulp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011657.2 | NP_035787.1 | 6 | 702 | ||
| GenBank mRNA | AB016963.1 | - | 6 | 645 | ||
| AF045582.1 | - | 6 | 655 | |||
| BC060068.1 | - | 6 | 632 | 6419643 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tulp3| Gene Symbol: Tulp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011657.2 | NP_035787.1 | 6 | 702 | ||
| GenBank mRNA | AB016963.1 | - | 6 | 645 | ||
| AF045582.1 | - | 6 | 655 | |||
| BC060068.1 | - | 6 | 632 | 6419643 | ||