siRNA Details
| Gene Symbol: Tulp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB016963.1 | - | 11 | 2801 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tulp3| Gene Symbol: Tulp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB016963.1 | - | 11 | 2801 | ||