siRNA Details
| Gene Symbol: Txn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011660.3 | NP_035790.1 | 5 | 741 | ||
| GenBank mRNA | AK007537.1 | - | 5 | 622 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Txn1| Gene Symbol: Txn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011660.3 | NP_035790.1 | 5 | 741 | ||
| GenBank mRNA | AK007537.1 | - | 5 | 622 | ||