siRNA Details
| Gene Symbol: Usp12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011669.3 | NP_035799.1 | 2 | 407 | ||
| GenBank mRNA | AF441835.1 | - | 1 | 68 | ||
| AK006739.1 | - | 2 | 457 | |||
| AK035629.1 | - | 2 | 248 | |||
| AK167153.1 | - | 2 | 247 | |||
| BC068136.1 | - | 2 | 407 | 30363916 | ||

Usp12
Genomic Map