siRNA Details
| Gene Symbol: Uchl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011670.2 | NP_035800.2 | 4, 5 | 504 | ||
| GenBank mRNA | AB025313.1 | - | 4, 5 | 355 | ||
| AF172334.1 | - | 4, 5 | 343 | |||
| AK004066.1 | - | 4, 5 | 381 | |||
| AK013729.1 | - | 4, 5 | 384 | |||
| AK170728.1 | - | 2, 3 | 342 | |||
| BC039177.1 | - | 4, 5 | 370 | 4506229 | ||
| CX241178.1 | - | 4, 5 | 504 | |||
| JX050004.1 | - | 2, 3 | 308 | |||

Uchl1
Genomic Map