siRNA Details
| Gene Symbol: Ufd1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011672.4 | NP_035802.3 | 4 | 338 | ||
| GenBank mRNA | AK010461.1 | - | 4 | 307 | ||
| AK012543.1 | - | 2 | 285 | |||
| AK088823.1 | - | 4 | 307 | |||
| AK147138.1 | - | 4 | 321 | |||
| AK169062.1 | - | 4 | 320 | |||
| BC006630.1 | - | 4 | 295 | 3708320 | ||
| CJ053815.1 | - | 4 | 307 | |||
| CJ085208.1 | - | 4 | 338 | |||
| U64445.1 | - | 4 | 323 | |||

Ufd1l
Genomic Map