siRNA Details
| Gene Symbol: Ufd1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011672.4 | NP_035802.3 | 12 | 1018 | ||
| GenBank mRNA | AK010461.1 | - | 12 | 987 | ||
| AK012543.1 | - | 10 | 962 | |||
| AK088823.1 | - | 12 | 987 | |||
| AK147138.1 | - | 12 | 1001 | |||
| AK169062.1 | - | 12 | 1000 | |||
| BC006630.1 | - | 12 | 975 | 3708320 | ||

Ufd1l
Genomic Map