siRNA Details
| Gene Symbol: Vmn1r51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011683.2 | NP_035813.2 | 3 | 2122 | ||
| GenBank mRNA | Y12725.1 | - | 2 | 2092 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn1r51| Gene Symbol: Vmn1r51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011683.2 | NP_035813.2 | 3 | 2122 | ||
| GenBank mRNA | Y12725.1 | - | 2 | 2092 | ||