siRNA Details
| Gene Symbol: Vill | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164567.1 | NP_001158039.1 | 4 | 248 | ||
| NM_011700.2 | NP_035830.2 | 4 | 248 | |||
| GenBank mRNA | AJ344341.1 | - | 4 | 236 | ||
| AK028229.1 | - | 4 | 249 | |||
| AK166823.1 | - | 3 | 480 | |||
| BB611096.1 | - | 4 | 249 | |||
| BC021808.1 | - | 4 | 224 | 3991439 | ||
| CN670511.1 | - | 4 | 224 | |||

Vill
Genomic Map