siRNA Details
| Gene Symbol: Vnn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011704.3 | NP_035834.2 | 5 | 1059 | ||
| GenBank mRNA | AJ132098.1 | - | 5 | 892 | ||
| AK002773.1 | - | 5 | 891 | |||
| AK145984.1 | - | 5 | 965 | |||
| AK149485.1 | - | 3 | 440 | |||
| AK167427.1 | - | 5 | 891 | |||
| BC019203.1 | - | 5 | 885 | 5041670 | ||

Vnn1
Genomic Map