siRNA Details
| Gene Symbol: Wnt8b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011720.3 | NP_035850.2 | 6 | 1474 | ||
| GenBank mRNA | AF130349.1 | - | 6 | 1476 | ||
| AK050992.1 | - | 6 | 1475 | |||
| AK169459.1 | - | 2 | 974 | |||
| BC119544.2 | - | 6 | 1407 | 40056929 | ||

Wnt8b
Genomic Map