siRNA Details
| Gene Symbol: Ercc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011729.2 | NP_035859.2 | 13 | 3019 | ||
| GenBank mRNA | AK137563.1 | - | 14 | 2851 | ||
| AK140825.1 | - | 13 | 2848 | |||
| AK160581.1 | - | 7 | 1893 | |||
| AK160645.1 | - | 11 | 2391 | |||
| AK164205.1 | - | 13 | 2837 | |||
| BC033413.1 | - | 13 | 2739 | |||
| BC053034.1 | - | 13 | 2829 | |||
| BC060659.1 | - | 12 | 3150 | |||
| BC092235.1 | - | 13 | 2810 | |||
| BC141070.1 | - | 13 | 2849 | 9055682 | ||
| D16306.1 | - | 13 | 2714 | |||
| U40796.1 | - | 13 | 2915 | |||

Ercc5
Genomic Map