siRNA Details
| Gene Symbol: Zfp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037665.2 | NP_001032754.1 | 4 | 666 | ||
| NM_011742.2 | NP_035872.2 | 4 | 624 | |||
| GenBank mRNA | AK134452.1 | - | 4 | 531 | ||
| AK147054.1 | - | 4 | 604 | |||
| AK153830.1 | - | 4 | 608 | |||
| BC025141.1 | - | 5 | 736 | |||
| BC055796.1 | - | 4 | 575 | 6412519 | ||
| BC062970.1 | - | 4 | 575 | 6412519 | ||
| X16493.1 | - | 4 | 733 | |||

Zfp1
Genomic Map