siRNA Details
| Gene Symbol: Slc30a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011773.3 | NP_035903.2 | 5, 6 | 955 | ||
| GenBank mRNA | AK144568.1 | - | 5, 6 | 939 | ||
| AK165796.1 | - | 5, 6 | 918 | |||
| U76007.1 | - | 5, 6 | 913 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc30a3| Gene Symbol: Slc30a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011773.3 | NP_035903.2 | 5, 6 | 955 | ||
| GenBank mRNA | AK144568.1 | - | 5, 6 | 939 | ||
| AK165796.1 | - | 5, 6 | 918 | |||
| U76007.1 | - | 5, 6 | 913 | |||