siRNA Details
| Gene Symbol: Slc30a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011774.3 | NP_035904.2 | 1, 2 | 635 | ||
| GenBank mRNA | AF003747.1 | - | 1, 2 | 436 | ||
| AK134245.1 | - | 1, 2 | 613 | |||
| BC117997.1 | - | 1, 2 | 493 | 40095367 | ||

Slc30a4
Genomic Map