siRNA Details
| Gene Symbol: Adam23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177600.1 | NP_001171071.1 | 4, 5 | 864 | ||
| NM_011780.2 | NP_035910.1 | 4, 5 | 864 | |||
| GenBank mRNA | AB009673.1 | - | 4, 5 | 636 | ||
| AK029301.1 | - | 2, 3 | 179 | |||
| AK034022.1 | - | 4, 5 | 859 | |||
| AK046677.1 | - | 4, 5 | 878 | |||
| AK083458.1 | - | 4, 5 | 835 | |||

Adam23
Genomic Map