siRNA Details
| Gene Symbol: Adamts5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011782.2 | NP_035912.2 | 1 | 1349 | ||
| GenBank mRNA | AF140673.1 | - | 1 | 508 | ||
| BC138619.1 | - | 1 | 533 | 8861639 | ||
| BC138620.1 | - | 1 | 533 | 8861640 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Adamts5| Gene Symbol: Adamts5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011782.2 | NP_035912.2 | 1 | 1349 | ||
| GenBank mRNA | AF140673.1 | - | 1 | 508 | ||
| BC138619.1 | - | 1 | 533 | 8861639 | ||
| BC138620.1 | - | 1 | 533 | 8861640 | ||