siRNA Details
| Gene Symbol: Arih2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011790.4 | NP_035920.1 | 13 | 1456 | ||
| GenBank mRNA | AF124664.1 | - | 12 | 1283 | ||
| AJ130975.1 | - | 12 | 1315 | |||
| AK011094.1 | - | 13 | 1463 | |||
| AK045012.1 | - | 1439 | ||||
| AK149936.1 | - | 13 | 1436 | |||
| AK167104.1 | - | 13 | 1413 | |||
| AK168055.1 | - | 13 | 1459 | |||
| BC051998.1 | - | 13 | 1456 | 6400287 | ||
| BC052422.1 | - | 13 | 1456 | 6400287 | ||

Arih2
Genomic Map