siRNA Details
Gene Symbol: Ets2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011809.3 | NP_035939.3 | 10 | 2265 | ||
GenBank mRNA | AK014602.2 | - | 10 | 2187 | ||
AK030537.1 | - | 6 | 1567 | |||
AK038001.1 | - | 1 | 2026 | |||
AK038348.1 | - | 10 | 2252 | |||
AK128933.1 | - | 11 | 2139 | |||
AK143687.1 | - | 10 | 2217 | |||
AK144974.1 | - | 10 | 2216 | |||
AK145852.1 | - | 10 | 2101 | |||
AK151315.1 | - | 10 | 2216 | |||
AK152750.1 | - | 10 | 2219 | |||
AK153339.1 | - | 10 | 2217 | |||
AK153478.1 | - | 10 | 2217 | |||
BC005486.1 | - | 10 | 2186 | 3486021 | ||
BC005504.1 | - | 10 | 2126 | 3490813 |
