siRNA Details
| Gene Symbol: Gadd45g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011817.2 | NP_035947.2 | 4 | 608 | ||
| GenBank mRNA | AB021884.1 | - | 4 | 605 | ||
| AF055638.1 | - | 4 | 596 | |||
| AK002237.1 | - | 4 | 606 | |||
| AK007410.1 | - | 4 | 611 | |||
| BC001989.1 | - | 4 | 589 | 3493618 | ||

Gadd45g
Genomic Map