siRNA Details
| Gene Symbol: Gpc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001079844.2 | NP_001073313.1 | 2 | 937 | ||
| NM_011821.3 | NP_035951.1 | 2 | 937 | |||
| GenBank mRNA | AF105268.1 | - | 3 | 660 | ||
| AK032866.1 | - | 2 | 358 | |||
| AK132531.1 | - | 2 | 674 | |||
| AK135442.1 | - | 2 | 919 | |||
| AK149312.1 | - | 2 | 704 | |||
| AK164668.1 | - | 2 | 939 | |||
| BC023448.1 | - | 2 | 351 | 5008374 | ||

Gpc6
Genomic Map