siRNA Details
| Gene Symbol: Grem1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011824.4 | NP_035954.1 | 2 | 413 | ||
| GenBank mRNA | AF045801.1 | - | 1 | 254 | ||
| AF108189.1 | - | 1 | 323 | |||
| AK135022.1 | - | 2 | 448 | |||
| AK157793.1 | - | 2 | 414 | |||
| AK164871.1 | - | 2 | 414 | |||
| BC015293.1 | - | 2 | 383 | 4216669 | ||

Grem1
Genomic Map