siRNA Details
| Gene Symbol: Insrr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011832.2 | NP_035962.2 | 12 | 2795 | ||
| GenBank mRNA | AB007135.1 | - | 12 | 2684 | ||
| BC137855.1 | - | 12 | 2652 | 8860874 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Insrr| Gene Symbol: Insrr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011832.2 | NP_035962.2 | 12 | 2795 | ||
| GenBank mRNA | AB007135.1 | - | 12 | 2684 | ||
| BC137855.1 | - | 12 | 2652 | 8860874 | ||