siRNA Details
| Gene Symbol: Mid2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011845.2 | NP_035975.1 | 8 | 1485 | ||
| GenBank mRNA | AF196480.1 | - | 8 | 1485 | ||
| Y18881.1 | - | 9 | 1840 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mid2| Gene Symbol: Mid2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011845.2 | NP_035975.1 | 8 | 1485 | ||
| GenBank mRNA | AF196480.1 | - | 8 | 1485 | ||
| Y18881.1 | - | 9 | 1840 | |||