siRNA Details
| Gene Symbol: Mmp17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011846.4 | NP_035976.3 | 10 | 3373 | ||
| GenBank mRNA | AK079221.1 | - | 3145 | |||
| AK143521.1 | - | 10 | 3374 | |||
| BC051917.1 | - | 10 | 3373 | 5716458 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mmp17| Gene Symbol: Mmp17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011846.4 | NP_035976.3 | 10 | 3373 | ||
| GenBank mRNA | AK079221.1 | - | 3145 | |||
| AK143521.1 | - | 10 | 3374 | |||
| BC051917.1 | - | 10 | 3373 | 5716458 | ||