siRNA Details
| Gene Symbol: Nek3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162947.1 | NP_001156419.1 | 9 | 910 | ||
| NM_011848.4 | NP_035978.2 | 10 | 968 | |||
| GenBank mRNA | AF093416.1 | - | 9 | 915 | ||
| AF099066.1 | - | 10 | 937 | |||
| BC005411.1 | - | 10 | 945 | 3590866 | ||
| CV560683.1 | - | 3 | 322 | |||

Nek3
Genomic Map