siRNA Details
Gene Symbol: Nlrp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF074018.1 | - | 1 | 44 | ||
AK087774.1 | - | 2 | 112 | |||
AY196361.1 | - | 1 | 44 | |||
AY196362.1 | - | 1 | 44 | |||
AY329484.1 | - | 1 | 44 | |||
AY329485.1 | - | 1 | 44 | |||
AY329486.1 | - | 1 | 44 | |||
AY329487.1 | - | 1 | 44 | |||
AY329488.1 | - | 1 | 44 | |||
AY329489.1 | - | 1 | 44 | |||
AY329490.1 | - | 1 | 44 | |||
AY329491.1 | - | 1 | 44 | |||
BC053384.1 | - | 2 | 111 | 30049556 |
